Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hil-8
Entrez Gene ID
188129
Gene Name
H15 domain-containing protein
Gene Aliases
CELE_T05E8.2
Chromosome Location
Chr.I: 5463117 - 5469639 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059202.5 NP_491603.2 2 - 3 358 76

View Full Product Details