Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ceh-12
Entrez Gene ID
172255
Gene Name
protein-coding
Gene Aliases
CELE_F33D11.4
Chromosome Location
Chr.I: 5840282 - 5841940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322676.1 NP_001309522.1 1 - 2 125 156
NM_059292.2 NP_491693.1 3 - 4 459 156

View Full Product Details