Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttll-11
Entrez Gene ID
177677
Gene Name
protein-coding
Gene Aliases
CELE_H23L24.3
Chromosome Location
Chr.IV: 8697736 - 8702701 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380378.1 NP_001367737.1 9 - 10 1273 67
NM_171405.7 NP_741471.2 11 - 12 1570 67

View Full Product Details