Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exc-5
Entrez Gene ID
177692
Gene Name
protein-coding
Gene Aliases
CELE_C33D9.1
Chromosome Location
Chr.IV: 8799081 - 8813670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027868.5 NP_001023039.1 14 - 15 2269 85
NM_001307099.3 NP_001294028.1 13 - 14 2068 85
NM_001307100.4 NP_001294029.1 15 - 16 2505 85
GenBank mRNA AB060647.1 - 14 - 15 2322 85
AF339059.1 - 14 - 15 2322 85

View Full Product Details