Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dlc-3
Entrez Gene ID
259606
Gene Name
protein-coding
Gene Aliases
CELE_Y10G11A.2
Chromosome Location
Chr.IV: 16432225 - 16437958 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083248.4 NP_001076717.1 2 - 3 256 110
NM_001083249.4 NP_001076718.1 2 - 3 343 110

View Full Product Details