Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srr-10
Entrez Gene ID
259826
Gene Name
Serpentine Receptor, class R
Gene Aliases
CELE_T05B11.6
Chromosome Location
Chr.V: 7735619 - 7737024 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171483.1 NP_741566.1 4 - 5 800 118

View Full Product Details