Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-24
Entrez Gene ID
179767
Gene Name
protein-coding
Gene Aliases
CELE_R90.5
Chromosome Location
Chr.V: 12882029 - 12885960 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269533.2 NP_001256462.1 5 - 6 432 75
NM_001269534.3 NP_001256463.1 4 - 5 360 75
NM_001269535.3 NP_001256464.1 3 - 4 210 75

View Full Product Details