Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35C1
Entrez Gene ID
179885
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C06B3.3
Chromosome Location
Chr.V: 13896663 - 13898496 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171550.5 NP_741638.1 1 - 2 193 79

View Full Product Details