Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-2
Entrez Gene ID
180570
Gene Name
protein-coding
Gene Aliases
CELE_T02C5.5
Chromosome Location
Chr.X: 2710920 - 2763526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348588.4 NP_001335561.1 5 - 6 857 151
NM_001348589.4 NP_001335563.1 5 - 6 857 151
NM_001348590.4 NP_001335560.1 5 - 6 857 151
NM_001348591.4 NP_001335562.1 5 - 6 857 151
NM_001348592.5 NP_001335571.1 5 - 6 1052 151
NM_001348593.4 NP_001335573.1 5 - 6 1052 151
NM_001348594.5 NP_001335570.1 5 - 6 1052 151
NM_001348595.4 NP_001335572.1 5 - 6 1052 151
NM_001348596.5 NP_001335516.1 3 - 4 584 151
NM_001348597.2 NP_001335515.1 3 - 4 584 151
NM_001368494.4 NP_001355464.1 11 - 12 1394 151
NM_001368495.4 NP_001355463.1 9 - 10 1238 151
NM_001368496.4 NP_001355462.1 9 - 10 1262 151
NM_001368497.4 NP_001355494.1 5 - 6 1161 151
NM_171638.3 NP_741732.2 5 - 6 1018 151
GenBank mRNA AY264781.1 - 5 - 6 1018 151

View Full Product Details