Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gnrr-4
Entrez Gene ID
180841
Gene Name
protein-coding
Gene Aliases
CELE_C41G11.4
Chromosome Location
Chr.X: 5738756 - 5743309 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373308.3 NP_001360691.1 11 - 12 1289 84
NM_171692.7 NP_741798.1 11 - 12 1297 84

View Full Product Details