Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pkc-2
Entrez Gene ID
181166
Gene Name
protein-coding
Gene Aliases
CELE_E01H11.1
Chromosome Location
Chr.X: 9364687 - 9386044 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029345.6 NP_001024516.1 12 - 13 1913 85
NM_001129650.5 NP_001123122.1 14 - 15 2258 85
NM_001373418.2 NP_001360727.1 13 - 14 2251 85

View Full Product Details