Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pkc-2
Entrez Gene ID
181166
Gene Name
protein-coding
Gene Aliases
CELE_E01H11.1
Chromosome Location
Chr.X: 9364687 - 9386044 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029345.6 NP_001024516.1 5 - 6 570 93
NM_001029346.4 NP_001024517.1 5 - 6 571 93
NM_001129650.5 NP_001123122.1 7 - 8 915 93
NM_001373417.3 NP_001360728.1 7 - 7 677 93
NM_001373418.2 NP_001360727.1 6 - 7 908 93
NM_001373419.3 NP_001359883.1 6 - 7 755 93

View Full Product Details