Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-58
Entrez Gene ID
181224
Gene Name
protein-coding
Gene Aliases
CELE_T06H11.1
Chromosome Location
Chr.X: 10104610 - 10133182 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171758.5 NP_741880.1 1 - 2 136 112

View Full Product Details