Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyc-1
Entrez Gene ID
181515
Gene Name
protein-coding
Gene Aliases
CELE_C33G3.1
Chromosome Location
Chr.X: 14049535 - 14063890 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029268.5 NP_001024439.1 7 - 8 721 88
NM_171796.5 NP_741925.2 3 - 4 564 88
GenBank mRNA AJ133838.1 - 3 - 4 289 88

View Full Product Details