Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-53
Entrez Gene ID
188702
Gene Name
protein-coding
Gene Aliases
CELE_T21F2.1
Chromosome Location
Chr.X: 16711681 - 16720670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375204.1 NP_001361941.1 11 - 12 1334 83
NM_171812.7 NP_741945.3 12 - 13 1518 83

View Full Product Details