Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wip-1
Entrez Gene ID
175681
Gene Name
protein-coding
Gene Aliases
CELE_R144.4
Chromosome Location
Chr.III: 5018639 - 5021120 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171863.4 NP_741123.2 4 - 5 552 100

View Full Product Details