Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-35
Entrez Gene ID
173800
Gene Name
protein-coding
Gene Aliases
CELE_F35D11.2
Chromosome Location
Chr.II: 4612989 - 4626106 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381396.1 NP_001367814.1 2 - 3 393 140
NM_001381397.2 NP_001367813.1 3 - 4 465 140

View Full Product Details