Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctns-1
Entrez Gene ID
174308
Gene Name
protein-coding
Gene Aliases
CELE_C41C4.7
Chromosome Location
Chr.II: 8134737 - 8136918 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182222.6 NP_872022.1 5 - 6 1096 120

View Full Product Details