Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lst-4
Entrez Gene ID
178361
Gene Name
protein-coding
Gene Aliases
CELE_Y37A1B.2
Chromosome Location
Chr.IV: 14056670 - 14069860 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276882.3 NP_001263811.1 7 - 8 1453 68
NM_001307068.3 NP_001293997.1 2 - 3 474 68
NM_001307159.3 NP_001294088.1 2 - 3 480 68
NM_001383241.1 5 - 6 1122 68
NM_001383242.1 6 - 7 1763 68
NM_182290.5 NP_872090.1 5 - 6 1120 68

View Full Product Details