Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin-40
Entrez Gene ID
178788
Gene Name
protein-coding
Gene Aliases
CELE_T27C4.4
Chromosome Location
Chr.V: 3704988 - 3712796 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330804.3 NP_001317834.1 3 - 4 2125 77
NM_001373016.2 NP_001360139.1 6 - 6 2554 77
NM_071630.5 NP_504031.1 5 - 6 2547 77
NM_071631.7 NP_504032.1 5 - 6 2554 77

View Full Product Details