Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpn-4
Entrez Gene ID
172996
Gene Name
Calponin-homology (CH) domain-containing protein
Gene Aliases
CELE_F49D11.8
Chromosome Location
Chr.I: 10924240 - 10925753 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060448.4 NP_492849.3 4 - 5 408 79

View Full Product Details