Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-17
Entrez Gene ID
173108
Gene Name
GLYcosylation related
Gene Aliases
CELE_T15D6.3
Chromosome Location
Chr.I: 12376904 - 12378674 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060733.2 NP_493134.2 5 - 6 728 150

View Full Product Details