Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-23
Entrez Gene ID
188194
Gene Name
Serpentine receptor class alpha-23
Gene Aliases
CELE_T06G6.1
Chromosome Location
Chr.I: 12703435 - 12704727 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060818.1 NP_493219.1 1 - 2 256 83

View Full Product Details