Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bath-34
- Entrez Gene ID
-
24104971
- Gene Name
- BTB and MATH domain-containing protein 34
- Gene Aliases
-
CELE_W02A11.5
- Chromosome Location
-
Chr.I: 12750036 - 12752163 on Build WBcel235
- Gene Symbol
- B0019.2
- Entrez Gene ID
-
173147
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_B0019.2
- Chromosome Location
-
Chr.I: 12751599 - 12782343 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060836.7 |
NP_493237.3 |
1 - 2 |
261 |
88 |
- Gene Symbol
- bath-35
- Entrez Gene ID
-
24104973
- Gene Name
- BTB domain-containing protein
- Gene Aliases
-
CELE_W02A11.8
- Chromosome Location
-
Chr.I: 12750034 - 12759577 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383738.1 |
|
3 - 4 |
994 |
88 |
View Full Product Details