Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gsk-3
Entrez Gene ID
173149
Gene Name
Glycogen synthase kinase-3
Gene Aliases
CELE_Y18D10A.5
Chromosome Location
Chr.I: 12819214 - 12825603 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383740.1 2 - 3 514 96

View Full Product Details