Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pbs-2
Entrez Gene ID
173168
Gene Name
Proteasome subunit beta
Gene Aliases
CELE_C47B2.4
Chromosome Location
Chr.I: 12967216 - 12968280 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060870.4 NP_493271.1 1 - 2 586 98

View Full Product Details