Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aex-6
Entrez Gene ID
173223
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y87G2A.4
Chromosome Location
Chr.I: 13543013 - 13545188 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060975.5 NP_493376.1 5 - 6 585 104
GenBank mRNA AB112930.1 - 5 - 6 581 104

View Full Product Details