Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lagr-1
Entrez Gene ID
189370
Gene Name
Probable ceramide synthase lagr-1
Gene Aliases
CELE_Y6B3B.10
Chromosome Location
Chr.I: 13724577 - 13732047 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061002.3 NP_493403.1 3 - 4 314 101

View Full Product Details