Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ero-1
Entrez Gene ID
173325
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8B.8
Chromosome Location
Chr.I: 14664449 - 14674841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377680.2 NP_001364667.1 4 - 5 790 103
NM_001377681.1 NP_001364626.1 5 - 6 685 103
NM_001377682.2 NP_001364632.1 3 - 4 266 103

View Full Product Details