Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ero-1
Entrez Gene ID
173325
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8B.8
Chromosome Location
Chr.I: 14664449 - 14674841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306232.3 NP_001293161.1 2 - 3 598 71
NM_001377680.2 NP_001364667.1 6 - 7 1389 71
NM_001377681.1 NP_001364626.1 7 - 8 1284 71
NM_001377682.2 NP_001364632.1 5 - 6 865 71

View Full Product Details