Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
let-49
Entrez Gene ID
173353
Gene Name
protein-coding
Gene Aliases
CELE_Y54E5B.3
Chromosome Location
Chr.I: 14820381 - 14821651 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061184.5 NP_493585.1 1 - 2 112 85

View Full Product Details