Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eva-1
Entrez Gene ID
173355
Gene Name
protein-coding
Gene Aliases
CELE_F32A7.3
Chromosome Location
Chr.I: 14829667 - 14834841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061189.6 NP_493590.2 1 - 2 140 92

View Full Product Details