Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eva-1
Entrez Gene ID
173355
Gene Name
protein-coding
Gene Aliases
CELE_F32A7.3
Chromosome Location
Chr.I: 14829667 - 14834841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322695.3 NP_001309519.1 3 - 3 253 82
NM_001381268.1 NP_001366909.1 2 - 3 981 82
NM_061189.6 NP_493590.2 2 - 3 244 82

View Full Product Details