Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dog-1
Entrez Gene ID
173370
Gene Name
Helicase ATP-binding domain-containing protein
Gene Aliases
CELE_F33H2.1
Chromosome Location
Chr.I: 15017283 - 15023017 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061217.6 NP_493618.1 11 - 12 2217 138

View Full Product Details