Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sqt-2
Entrez Gene ID
173382
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_C01B12.1
Chromosome Location
Chr.II: 23325 - 24458 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061234.6 NP_493635.1 1 - 2 183 68

View Full Product Details