Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nol-2
Entrez Gene ID
173437
Gene Name
protein-coding
Gene Aliases
CELE_W07E6.1
Chromosome Location
Chr.II: 471240 - 475940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061341.6 NP_493742.1 3 - 4 729 73

View Full Product Details