Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mab-9
Entrez Gene ID
173441
Gene Name
T-box protein 12
Gene Aliases
CELE_T27A1.6
Chromosome Location
Chr.II: 516014 - 521635 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061349.8 NP_493750.1 3 - 4 454 127
GenBank mRNA AJ252168.1 - 3 - 4 366 127

View Full Product Details