Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mab-9
Entrez Gene ID
173441
Gene Name
T-box protein 12
Gene Aliases
CELE_T27A1.6
Chromosome Location
Chr.II: 516014 - 521635 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061349.8 NP_493750.1 4 - 5 543 121
GenBank mRNA AJ252168.1 - 4 - 5 455 121

View Full Product Details