Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-45
Entrez Gene ID
191917
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_K07E8.9
Chromosome Location
Chr.II: 659828 - 661796 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061381.4 NP_493782.2 1 - 2 203 116

View Full Product Details