Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acdh-9
Entrez Gene ID
173466
Gene Name
Acyl CoA DeHydrogenase
Gene Aliases
CELE_F28A10.6
Chromosome Location
Chr.II: 846864 - 848603 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061431.4 NP_493832.1 4 - 5 557 79

View Full Product Details