Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pod-2
Entrez Gene ID
173500
Gene Name
protein-coding
Gene Aliases
CELE_W09B6.1
Chromosome Location
Chr.II: 1135534 - 1152705 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267098.3 NP_001254027.1 8 - 9 2711 58
NM_001381323.1 NP_001368018.1 9 - 10 2849 58

View Full Product Details