Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-47
Entrez Gene ID
173532
Gene Name
BTB and MATH domain-containing protein 47
Gene Aliases
CELE_T07H3.1
Chromosome Location
Chr.II: 1601789 - 1604621 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061605.4 NP_494006.2 7 - 8 1973 99

View Full Product Details