Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btb-8
Entrez Gene ID
173540
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F45C12.6
Chromosome Location
Chr.II: 1722770 - 1724186 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061656.4 NP_494057.1 3 - 4 616 89

View Full Product Details