Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-95
Entrez Gene ID
183723
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_C52E2.1
Chromosome Location
Chr.II: 1854394 - 1855365 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061696.4 NP_494097.1 1 - 2 84 77

View Full Product Details