Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-15
Entrez Gene ID
182659
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_C16C4.5
Chromosome Location
Chr.II: 1866381 - 1867894 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061700.3 NP_494101.2 1 - 2 130 168

View Full Product Details