Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-20
Entrez Gene ID
181824
Gene Name
BTB and MATH domain containing
Gene Aliases
CELE_B0047.1
Chromosome Location
Chr.II: 2040276 - 2042631 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038232.4 NP_001033321.1 5 - 6 998 96

View Full Product Details