Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsr-1
Entrez Gene ID
173601
Gene Name
protein-coding
Gene Aliases
CELE_F53G2.6
Chromosome Location
Chr.II: 2460842 - 2473370 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383801.1 2 - 3 241 67

View Full Product Details