Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-60
Entrez Gene ID
191924
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_Y47G7B.3
Chromosome Location
Chr.II: 2701389 - 2702857 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061936.1 NP_494337.1 3 - 4 498 69

View Full Product Details