Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nep-2
- Entrez Gene ID
-
188090
- Gene Name
- Neprilysin-2
- Gene Aliases
-
CELE_T05A8.4
- Chromosome Location
-
Chr.II: 2716555 - 2725752 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061942.3 |
NP_494343.2 |
16 - 17 |
2143 |
125 |
- Gene Symbol
- T05A8.3
- Entrez Gene ID
-
173615
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_T05A8.3
- Chromosome Location
-
Chr.II: 2714640 - 2725757 on Build WBcel235
View Full Product Details