Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-88
Entrez Gene ID
173641
Gene Name
protein-coding
Gene Aliases
CELE_K08A2.5
Chromosome Location
Chr.II: 2978408 - 2987023 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383805.1 6 - 7 1176 83

View Full Product Details